missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Glass Screw Neck Vial, 10-425 Thread

Wide opening enables easy filling with viscous materials.

Brand:  Fisherbrand™ 10 09 1196

28.57 EUR valid until 2025-03-30
Use promo code "24134" to get your promotional price.


 View more versions of this product

Product Code. 10521593

  • 35.00 € / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Wide opening enables easy filling with viscous materials
  • Any combination of 1.5ml Screw Neck Vial 10-425 with 10mm PP Screw Seals can be obtained as a 2in1 KIT
  • Closed top Seals and replacement septa are available
  • Broad range of Micro-Inserts
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Clear
Glass
GC, HPLC
CleanPack, Polypropylene Box
1.5 mL
32 mm
ND10
11.6 mm
2 mL
Screw
<120 μL
Flat Bottom
100
Yes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.