missing translation for 'onlineSavingsMsg'
Learn More

Glutathione, 98%, for analysis, reduced, Thermo Scientific Chemicals

32.76 € - 2810.99 €

Chemical Identifiers

CAS 70-18-8
Molecular Formula C10H17N3O6S
Molecular Weight (g/mol) 307.32
MDL Number MFCD00065939
InChI Key RWSXRVCMGQZWBV-WDSKDSINSA-N
Synonym glutathione, l-glutathione, glutathion, glutathione-sh, glutinal, isethion, tathion, reduced glutathione, deltathione, neuthion
PubChem CID 124886
ChEBI CHEBI:16856
IUPAC Name (2S)-2-amino-5-[[(2R)-1-(carboxymethylamino)-1-oxo-3-sulfanylpropan-2-yl]amino]-5-oxopentanoic acid
SMILES C(CC(=O)NC(CS)C(=O)NCC(=O)O)C(C(=O)O)N
View More Specs

Products 5
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10346940
View Documents
Thermo Scientific
120000010
1 g
32.76 €
1g
Please to purchase this item. Need a web account? Register with us today!
10021090
View Documents
Thermo Scientific Acros
120000050
5 g
59.70 €
5g
Please to purchase this item. Need a web account? Register with us today!
10418840
View Documents
Thermo Scientific Acros
120000250
25 g
238.19 €
25g
Please to purchase this item. Need a web account? Register with us today!
10628343
View Documents
Thermo Scientific Acros
120001000
100 g
714.56 €
100g
Please to purchase this item. Need a web account? Register with us today!
11975721
View Documents
Thermo Scientific Acros
120005000
500 g
2810.99 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

70-18-8
307.32
RWSXRVCMGQZWBV-WDSKDSINSA-N
124886
(2S)-2-amino-5-[[(2R)-1-(carboxymethylamino)-1-oxo-3-sulfanylpropan-2-yl]amino]-5-oxopentanoic acid
C10H17N3O6S
MFCD00065939
glutathione, l-glutathione, glutathion, glutathione-sh, glutinal, isethion, tathion, reduced glutathione, deltathione, neuthion
CHEBI:16856
C(CC(=O)NC(CS)C(=O)NCC(=O)O)C(C(=O)O)N

Specifications

70-18-8
White
Authentic
Glass bottle
MFCD00065939
15,4511
− 16.50
Solubility in water: very soluble. Other solubilities: freely soluble in dilute alcohol and dimethyl-, formamide, almost insoluble in ethanol
C(CC(=O)NC(CS)C(=O)NCC(=O)O)C(C(=O)O)N
307.32
CHEBI:16856
98%
Crystalline Powder
182.0°C to 192.0°C
0.5% max. (105°C, 3 hrs)
97.5% min. (Iodimetry)
C10H17N3O6S
04,II,931
− 16.50 (20.00°C c=2,H2O)
glutathione, l-glutathione, glutathion, glutathione-sh, glutinal, isethion, tathion, reduced glutathione, deltathione, neuthion
RWSXRVCMGQZWBV-WDSKDSINSA-N
(2S)-2-amino-5-[[(2R)-1-(carboxymethylamino)-1-oxo-3-sulfanylpropan-2-yl]amino]-5-oxopentanoic acid
124886
307.32
Analysis
Glutathione, For analysis, Reduced
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 200-725-4

Product Identifier
  • Glutathione reduced

SDS
Documents

Documents

Certificates

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.