missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Glutathione Sepharose™ 4 Fast Flow Media
Single-step purification of GST-tagged proteins and other glutathione S-transferase or glutathione-dependent proteins. Glutathione Sepharose 4 Fast Flow Media is used for the purification of GST-tagged proteins from bacterial lysates and other S-transferases or glutathione-dependent proteins.
740.00 € - 8632.00 €
Specifications
Binding Capacity | >10 mg/mL |
---|---|
Description | Glutathione Sepharose 4 Fast Flow is an affinity resin especially suited for purification of GST-tagged proteins in a batch mode or when scale up is needed. |
Flow Rate | <450 cm/hr. |
Kit Contents | 10 Carbon Spacer Arm, 4% Agarose, 90 μm Particle Size |
Matrix | Highly cross-linked 4% agarose |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11595015
|
Cytiva
17-5132-02 |
100 mL |
2200.00 €
100mL |
Estimated Shipment: 07-10-2024
Log in to see stock. |
|||||
11585015
|
Cytiva
17-5132-01 |
25 mL |
740.00 €
25mL |
Estimated Shipment: 07-10-2024
Log in to see stock. |
|||||
11505025
|
Cytiva
17-5132-03 |
500 mL |
8632.00 €
500mL |
Estimated Shipment: 08-10-2024
Log in to see stock. |
|||||
Description
- Glutathione coupled to Sepharose 4 Fast Flow via 10-carbon spacer arm
- Protein binding capacity: ≈10mg recombinant GST/mL media (flowrate dependent)
- Binding capacity and high flow properties allow scale-up
- Mild elution conditions preserve protein antigenicity and function
- Stable in all commonly used aqueous buffers
- pH stability: 3 to 12
Specifications
>10 mg/mL | |
<450 cm/hr. | |
Highly cross-linked 4% agarose | |
4% Highly Cross-linked Agarose | |
base matrix 150-250 cm/h, 100 kPa, XK 50/60 column, bed height 25 cm | |
Chromatography Media |
Glutathione Sepharose 4 Fast Flow is an affinity resin especially suited for purification of GST-tagged proteins in a batch mode or when scale up is needed. | |
10 Carbon Spacer Arm, 4% Agarose, 90 μm Particle Size | |
3 to 12 | |
4°C to 30°C, 20% Ethanol | |
90 μm | |
Glutathione and 10-carbon linker arm |