missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Glutathione Sepharose™ High Performance Media

For high resolution purification of Glutathione S transferase (GST) tagged proteins

Brand:  Cytiva 17-5279-01


 View more versions of this product

Product Code. 10032949

  • 856.00 € / 25mL
Estimated Shipment: 14-10-2024
to see stock.
Add to basket
This item is not returnable. View return policy

Description

Description

  • One-step purification of GST-tagged proteins and other glutathione S-transferases or glutathione-dependent proteins
  • GST-tagged proteins are eluted under mild, nondenaturing conditions using reduced glutathione
  • Mild purification process preserves protein antigenicity and function
  • Glutathione ligand is coupled to 6% agarose matrix

TRUSTED_SUSTAINABILITY
Specifications

Specifications

>Approx. 10 mg GST-tagged protein/mL resin
<600 cm/hr.
Highly crossed-linked agarose, 6%
6% Highly Cross-linked Agarose
34 μm
Chromatography Media
Glutathione Sepharose High Performance is an affinity resin recommended for purifying glutathione S-transferase (GST)-tagged proteins when high resolution is the priority.
Binding capacity 10mg recombinant GST/mL medium, Highly crosslinked Agarose, 6%
3 to 12
4°C to 30°C, 20% Ethanol
25 mL
Glutathione
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Glutathione Sepharose High Performance, 25 ml

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers