missing translation for 'onlineSavingsMsg'
Learn More

Glycerol, 99+%, extra pure, Thermo Scientific Chemicals

31.36 € - 386.93 €

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglynShow MoreShow Less
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10562524
SDS View Documents
Thermo Scientific Acros
158922500
250 mL Glass bottle
31.36 €
250mL
Please to purchase this item. Need a web account? Register with us today!
10296200
SDS View accessory products View Documents
Thermo Scientific Acros
158920010
1 L Glass Bottle
91.35 €
1 L
Please to purchase this item. Need a web account? Register with us today!
10152970
SDS View Documents
Thermo Scientific Acros
158920025
2.5 L Glass Bottle
152.76 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
10648143
SDS View Documents
Thermo Scientific Acros
158920100
10 L Plastic drum
386.93 €
10 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 200-289-5

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Glycerol, 99+%, extra pure, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.