Learn More
Glycine hydrochloride, 98%, Thermo Scientific Chemicals
Description
Glycine hydrochloride is an inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Glycine hydrochloride is an inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors.
Solubility
Soluble in water (>100 g/L).
Notes
Hygroscopic. Store away from strong oxidizing agents, water/ moisture.

Specifications
Specifications
Melting Point | 175°C to 178°C |
Beilstein | 3909617 |
Merck Index | 14,4491 |
Solubility Information | Soluble in water (>100g/L). |
Quantity | 100 g |
Odor | Odorless |
Formula Weight | 111.53 |
Sensitivity | Hygroscopic |
Percent Purity | 98% |
Chemical Name or Material | Glycine hydrochloride |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Q07L024 | Certificate of Analysis | 08/04/2025 | A12162.0E,A12162.36,A12162.22 |
T11K007 | Certificate of Analysis | 26/07/2024 | A12162.0E,A12162.36,A12162.22 |
R01K010 | Certificate of Analysis | 08/05/2024 | A12162.0E,A12162.36,A12162.22 |
M09K001 | Certificate of Analysis | 17/01/2024 | A12162.0E,A12162.36,A12162.22 |
U31H039 | Certificate of Analysis | 23/12/2022 | A12162.0E,A12162.36,A12162.22 |
Safety and Handling

- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.