Learn More
Motic™ Bifurcated Gooseneck Light Guide
Half-meter long light guide allows a user to direct light source where it's needed. Motic™ Bifurcated Gooseneck Light Guide is for use with the Motic MLC-150 illuminator.
Brand: Motic™ 1101002000142
Description
- Bifurcated gooseneck style light guide is positionable, holding the angle of illumination required for observations
- Rugged fiber optic light guide provides cool and uniform illumination, and comes with a PVC coated, stainless steel sheathing
- 0.5 Meter length
- Use with MLC-150 halogen cold light illuminator
Specifications
Bifurcated Gooseneck | |
Halogen | |
Motic MLC-150 cold light illuminator (11-776-576) | |
0.5 m |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
Motic™ Bifurcated Gooseneck Light Guide > Gooseneck bifurcated light guide
Spot an opportunity for improvement?Share a Content Correction