missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCHOTT AG Lighting and Imaging™ Gooseneck Light Guide
Fiber optic flex and stay light guide in a black lacquered semi-rigid gooseneck tubing.
Brand: SCHOTT AG Lighting and Imaging™ 154202
This item is not returnable.
View return policy
Description
- Light guide with eco-friendly Puravis™ fiber optics.
- Precise positioning offers complete control of light placement.
- Gooseneck branches are positioned in opposite directions to create near shadow-free illumination.
- Re-adjust with ease.
- For use with cold light source KL1600LED, KL2500LED or KL1500HAL.
Specifications
Gooseneck Double Light Guide | |
11 mm | |
4.5 mm Fiber Bundle Diameter | |
Cold Light Sources KL1600LED, KL2500LED or KL1500HAL | |
600 mm | |
Optical glass fibers |
0.4 in. | |
4.5 mm | |
Stereo Microscopy Illumination | |
23.6 in. | |
600 mm |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 7
Videos
-
Fiber-optic-light-guides-for-incident-illumination-video_ENG
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SCHOTT AG Lighting and Imaging™ Gooseneck Light Guide > Gooseneck Light Guide; Type: Double Gooseneck; Fiber Diameter: 4.5 mm; Length: 600 mm
Spot an opportunity for improvement?Share a Content Correction