missing translation for 'onlineSavingsMsg'
Learn More

Gosselin™ Conical Base Polypropylene Test Tubes

Compatible with standard rotors. Gosselin™ Conical Base Polypropylene Test Tubes come with green screw cap.

Brand:  Gosselin™ TC50-04

 View more versions of this product

Product Code. 11763274

  • 150.40 € / Pack of 500
Estimated Shipment: 25-07-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Sterile
  • Centrifuge tubes
  • With detachable sterility certificate
TRUSTED_SUSTAINABILITY
Specifications

Specifications

EU 10/2011, ISO 11137
Natural
Polypropylene
500
Sterile
Yes
Yes
9600 x g
Green
50 mL
Centrifugation
50
Conical
29.2 mm
Conical Tube
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gosselin™ Conical Base Polypropylene Test Tubes > Sterile; Polypropylene; With Green Screw cap

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.