Learn More
Gosselin™ Polystrene Petri Dish
Crystal polystyrene dishes, with optimal flatness.
Brand:  Gosselin BP50-02
 View more versions of this product
Product Code. 11788573
  • 266.89 € / Pack of 1620
Estimated Shipment: 02-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Appropriate for aseptic production
  • Sterilized by irradiation
  • Flat bottom
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CE, IVD, EC N°1935/2004, ISO 11137
Polystyrene
Sterile
1
1620 Carton
14.2 mm
Natural
Round
Petri Dish
For use in clinical, environment, industries, microbiology
0
55 mm