missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Sterile Polyethylene Blender Bags

Universal bags for homogeneous suspension

Brand:  Corning™ FBAG-04

 View more versions of this product

Product Code. 11777919

  • 922.01 € / Pack of 1500
Estimated Shipment: 26-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Polyethylene (PE) bags
  • 80 to 3500 mL
  • With or without filter
  • Traceability on each filter bag
  • Sterile

Order Info

Use bag clip, Cat. No. 07-201-979, to close bags between sample preparation steps.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

With full filter
Natural
Sterile
500/Pk.
300 mm
400 mL
Polyethylene
1500/Cs.
50/Pk.
190 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.