Learn More
A&D Weighing GP Series Precision Balances Internal Calibration Class II Approved
GREENER_CHOICE
With capacities from 12 to 102 kg, these high precision industrial balances have a large weighing pan, and also feature IP65 dust and waterproof protection. SmartRange feature for improved precision, regardless of tare container weight.
Brand:  A&D Weighing GP-30K-EC
 View more versions of this product
Product Code. 16601132
  • 4655.00 € / Each
Explore more special offers
Description
Description
  • EC Type approved
  • Swing arm design or separate display and platform design
  • IP65 protected
  • Durable aluminum alloy & stainless steel construction
  • Easy calibration with internal calibration mass
  • SHS response speed of only 1.5 seconds
  • GLP, GMP and ISO compliant
  • RS-232C interface
  • Comparator output and analog output (optional)
  • Large Vacuum Fluorescent display
  • Data memory function
  • Counting function with ACAI (Automatic Counting Accuracy Improvement)
  • Smart range function (GP-32K/102K/32KS)
  • Comparator function
  • Underhook weighing for thermal and magnetic samples as well as for density calculation
  • Animal weighing function
Specifications
Specifications
31 kg
0.1 g
Industrial Precision Balance
VFD
1.5 seconds
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
A&D Weighing GP Series Precision Balances Internal Calibration Class II Approved > 31 kg x 0.1 g