Learn More
Grace™ Reveleris™ SRC C18 Cartridges
Ideal for speed, resolution and capacity
Brand: Grace™ 145152103
This item is not returnable.
View return policy
Description
- Withstand higher pressures (up to 200psig) and deliver greater loading capacity
- Superior, high load silica for excellent resolution
- Optimally packed to give high plate count
- Tighter particle size distribution and minimal fines keep backpressure low
- RFID tag streamlines Reveleris system (sold separately) operation by auto-loading cartridge specifications
- Compatible with other popular flash systems
Specifications
C18 | |
12 g | |
200 psi |
Reveleris SRC C18 Cartridge | |
For Speed, Resolution and Capacity, Conventional AppliCatIons, Polar Compounds and Popular Flash Systems |