Learn More
Grace™ Reveleris™ SRC C18 Cartridges
Ideal for speed, resolution and capacity
265.18 € - 537.03 €
Specifications
Material C18
For Use With (Application) For Speed, Resolution and Capacity, Conventional AppliCatIons, Polar Compounds and Popular Flash Systems
Max. Pressure 200 psi
View More Specs
Products 3
Product Code Brand Bed Weight (Metric) Type Price Quantity & Availability  
Product Code Brand Bed Weight (Metric) Type Price Quantity & Availability  
11353581
View Documents
Grace™
145152105
80 g Bonded Flash Cartridge This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11333581
View Documents
Grace™
145152103
12 g Reveleris SRC C18 Cartridge This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11393581
View Documents
Grace™
145152991
120 g Reveleris SRC C18 Cartridge This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Withstand higher pressures (up to 200psig) and deliver greater loading capacity
  • Superior, high load silica for excellent resolution
  • Optimally packed to give high plate count
  • Tighter particle size distribution and minimal fines keep backpressure low
  • RFID tag streamlines Reveleris system (sold separately) operation by auto-loading cartridge specifications
  • Compatible with other popular flash systems
Specifications
Specifications
C18
For Speed, Resolution and Capacity, Conventional AppliCatIons, Polar Compounds and Popular Flash Systems
200 psi