Learn More
Thermo Scientific™ Grade 25 Sapphire Spheres, Dia.: 2mm
Brand: Thermo Scientific™ 042063.KN
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications
Sapphire Sphere | |
2 mm | |
Grade 25 | |
100 Pieces |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.