missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Grade 604 Surface Protection Paper
Designed to protect laboratory surfaces against hazardous spills. Fisherbrand™ Grade 604 Surface Protection Paper guards against damage from acids, toxic, corrosive and radioactive fluids and impacts.
66.45 € - 169.52 €
Specifications
Product Type | Surface Protection Paper |
---|---|
Material | Grade 604 Paper |
Backing Material | Polyethylene |
Color | White |
Thickness | 0.25mm |
Product Code | Brand | Dimensions (L x W) | Packaging Type | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Packaging Type | Price | Quantity & Availability | ||||||
11710005
|
Fisherbrand™
0604R00022 |
50 m x 6 cm | Roll | N/A | N/A | N/A | |||||
11715375
|
Fisherbrand™
0604F00021 |
520 x 420 mm | Sheet | N/A | N/A | N/A | |||||
11728742
|
Fisherbrand™
0604F00022 |
570 x 460 mm | Sheet |
108.00 €
Pack of 100 |
In Stock
Log in to see stock. ![]() |
||||||
11758722
|
Fisherbrand™
0604R00021 |
50 m x 46 cm | Roll |
169.52 €
Each |
Estimated Shipment: 04-06-2025
Log in to see stock. |
||||||
Description
- Available in sheet or roll form
- Polyethylene backed 125g/m2 absorbent paper quickly soaks up spills and prevents flow through to work surface
Specifications
Surface Protection Paper | |
Polyethylene | |
0.25mm |
Grade 604 Paper | |
White |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Grade 604 Surface Protection Paper