missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Grade 608 Joseph Paper
Fisherbrand™ Grade 608 Joseph Paper is very absorbent and no fluffy.
Brand: Fisherbrand™ 0608M00002
This item is not returnable.
View return policy
Description
- Thin paper 25g/m2
- very absorbent and no fluffy
Specifications
500 mm | |
500 x 350 mm | |
Cleaning and drying of, Glass containers, tubes, flasks, microscopic plates, bottles in laboratories and hospitals | |
608 |
350 mm | |
Folded sheet | |
40 Pk. |