Learn More
Fisherbrand™ Grade 7 Cellulose Fiber and Cotton Linter Extraction Thimble
Suitable for all extractors systems.
Brand:  Fisherbrand™ 0007N00040
90.86 EUR valid until 2025-07-13
Use promo code "24135" to get your promotional price.
 View more versions of this product
Product Code. 11724053
  • 122.72 € / Pack of 25
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • 100% High pure cotton linters fibers
  • Neutral and fat free
  • High mechanical strength
  • Excellent retention capacity
Applications:
  • Extraction of sulphur
  • Extraction of the content of free lipids in food
  • Determination of dust and tar in industrial air and gases
  • Lacquer, binder analysis in paint colours
  • Control of emission smokes in industrial chimneys
  • Gravimetric analysis of dust in gases
Specifications
Specifications
33 mm
Extraction of Sulfur in gaseous masses, determination of dust and tar in industrial air and gases
Suitable for all extractors systems
Extraction Thimble
High Purity Cellulose Fibers and Cotton Linters
7
80 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Grade 7 Cellulose Fiber and Cotton Linter Extraction Thimble > Inside Diameter: 33mm; Length: 80mm