missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Grade 7 Cellulose Fiber and Cotton Linter Extraction Thimble
Suitable for all extractors systems.
54.75 € - 381.00 €
Specifications
Material | High Purity Cellulose Fibers and Cotton Linters |
---|---|
For Use With (Application) | Extraction of Sulfur in gaseous masses, determination of dust and tar in industrial air and gases |
Grade | 7 |
For Use With (Equipment) | Suitable for all extractors systems |
Product Type | Extraction Thimble |
Product Code | Brand | Diameter (Metric) Inside | Length (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inside | Length (Metric) | Price | Quantity & Availability | ||||||
11764526
|
Fisherbrand™
0007N00021 |
16 mm | 100 mm | N/A | N/A | N/A | |||||
11734536
|
Fisherbrand™
0007N00122 |
19 mm | 40 mm | N/A | N/A | N/A | |||||
11724043
|
Fisherbrand™
0007N00022 |
19 mm | 90 mm |
54.75 €
Pack of 25 |
Estimated Shipment: 23-05-2025
Log in to see stock. |
||||||
11774526
|
Fisherbrand™
0007N00023 |
22 mm | 60 mm | N/A | N/A | N/A | |||||
11754043
|
Fisherbrand™
0007N00121 |
25 mm | 100 mm |
69.35 €
Pack of 25 |
Estimated Shipment: 03-06-2025
Log in to see stock. |
||||||
11734043
|
Fisherbrand™
0007N00024 |
22 mm | 80 mm |
62.20 €
Pack of 25 |
In Stock
Log in to see stock. ![]() |
||||||
11774043
|
Fisherbrand™
0007N00026 |
26 mm | 60 mm |
65.90 €
Pack of 25 |
Estimated Shipment: 15-05-2025
Log in to see stock. |
||||||
11744043
|
Fisherbrand™
0007N00025 |
25 mm | 80 mm |
66.45 €
Pack of 25 |
In Stock
Log in to see stock. ![]() |
||||||
11784526
|
Fisherbrand™
0007N00027 |
27 mm | 80 mm | N/A | N/A | N/A | |||||
11794043
|
Fisherbrand™
0007N00029 |
28 mm | 100 mm |
62.60 €
Pack of 25 |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
11542103
|
Fisherbrand™
0007N00128 |
28 mm | 120 mm | N/A | N/A | N/A | |||||
11784043
|
Fisherbrand™
0007N00028 |
28 mm | 80 mm |
97.95 €
Pack of 25 |
Estimated Shipment: 15-05-2025
Log in to see stock. |
||||||
11714053
|
Fisherbrand™
0007N00030 |
30 mm | 100 mm |
73.65 €
Pack of 25 |
Estimated Shipment: 09-07-2025
Log in to see stock. |
||||||
11724536
|
Fisherbrand™
0007N00120 |
30 mm | 150 mm | N/A | N/A | N/A | |||||
11704053
|
Fisherbrand™
0007N00031 |
30 mm | 80 mm | N/A | N/A | N/A | |||||
11734053
|
Fisherbrand™
0007N00041 |
33 mm | 100 mm |
63.50 €
Pack of 25 |
Estimated Shipment: 03-06-2025
Log in to see stock. |
||||||
11744536
|
Fisherbrand™
0007N00123 |
33 mm | 118 mm |
78.35 €
Pack of 25 |
Estimated Shipment: 11-06-2025
Log in to see stock. |
||||||
11724053
|
Fisherbrand™
0007N00040 |
33 mm | 80 mm |
122.72 €
Pack of 25 |
In Stock
Log in to see stock. ![]() |
||||||
11714536
|
Fisherbrand™
0007N00052 |
33 mm | 94 mm | N/A | N/A | N/A | |||||
11794526
|
Fisherbrand™
0007N00033 |
34 mm | 130 mm | N/A | N/A | N/A | |||||
11764536
|
Fisherbrand™
0007N00125 |
35 mm | 150 mm | N/A | N/A | N/A | |||||
11744053
|
Fisherbrand™
0007N00034 |
37 mm | 130 mm |
75.85 €
Pack of 25 |
Estimated Shipment: 20-05-2025
Log in to see stock. |
||||||
11754053
|
Fisherbrand™
0007N00035 |
41 mm | 123 mm |
89.95 €
Pack of 25 |
Estimated Shipment: 15-05-2025
Log in to see stock. |
||||||
11704536
|
Fisherbrand™
0007N00042 |
41 mm | 150 mm | N/A | N/A | N/A | |||||
11754536
|
Fisherbrand™
0007N00124 |
43 mm | 123 mm |
125.00 €
Pack of 25 |
Estimated Shipment: 04-06-2025
Log in to see stock. |
||||||
11774536
|
Fisherbrand™
0007N00130 |
58 mm | 170 mm | N/A | N/A | N/A | |||||
11784536
|
Fisherbrand™
0007N00131 |
70 mm | 240 mm | N/A | N/A | N/A | |||||
11794536
|
Fisherbrand™
0007N00132 |
80 mm | 200 mm | N/A | N/A | N/A | |||||
11704546
|
Fisherbrand™
0007N00133 |
80 mm | 250 mm | N/A | N/A | N/A | |||||
Description
- 100% High pure cotton linters fibers
- Neutral and fat free
- High mechanical strength
- Excellent retention capacity
Applications:
- Extraction of sulphur
- Extraction of the content of free lipids in food
- Determination of dust and tar in industrial air and gases
- Lacquer, binder analysis in paint colours
- Control of emission smokes in industrial chimneys
- Gravimetric analysis of dust in gases
Specifications
High Purity Cellulose Fibers and Cotton Linters | |
7 | |
Extraction Thimble |
Extraction of Sulfur in gaseous masses, determination of dust and tar in industrial air and gases | |
Suitable for all extractors systems |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Grade 7 Cellulose Fiber and Cotton Linter Extraction Thimble