missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Grade 925 Sterilization Paper

Used as wrap for medical clothes.

Brand:  Fisherbrand™ 0925F00029

 View more versions of this product

Product Code. 11700575

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Creped paper
  • Weight: 60g/mm2

Applications:

  • Used in steam gas and gamma-ray sterilization
Specifications

Specifications

Autoclavable
White
65 cm
60 g/m2
Paper
65 cm
EN 868-2
Wrapping of medical clothes, surgical instruments and others articles used in steam, gas and gamma-ray sterilization
Sterilization Paper
925
500 Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.