missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Grade 925 Sterilization Paper

Used as wrap for medical clothes.

127.00 € - 251.00 €

Specifications

Autoclavable Autoclavable
Certifications/Compliance EN 868-2
Color White
For Use With (Application) Wrapping of medical clothes, surgical instruments and others articles used in steam, gas and gamma-ray sterilization
Product Type Sterilization Paper
View More Specs

Products 7
Product Code Brand Width (Metric) Exterior DimensionsLxW Price Quantity & Availability  
Product Code Brand Width (Metric) Exterior DimensionsLxW Price Quantity & Availability  
11784576
View Documents
Fisherbrand™
0925F00026
100 cm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11754516
View Documents
Fisherbrand™
0925F00021
35 cm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11790565
View Documents
Fisherbrand™
0925F00023
60 cm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11780565
View Documents
Fisherbrand™
0925F00022
50 cm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11700575
View Documents
Fisherbrand™
0925F00029
65 cm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11764576
View Documents
Fisherbrand™
0925F00024
75 cm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11774576
View Documents
Fisherbrand™
0925F00025
900 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Creped paper
  • Weight: 60g/mm2

Applications:

  • Used in steam gas and gamma-ray sterilization
Specifications

Specifications

Autoclavable
White
Sterilization Paper
925
EN 868-2
Wrapping of medical clothes, surgical instruments and others articles used in steam, gas and gamma-ray sterilization
60 g/m2
Paper
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.