Learn More
Hirschmann™ Graduated Pipettes with Piston
Provides volumetric accuracy for your pipetting needs. Hirschmann Instruments™ Graduated Pipettes with Piston come with amber stain graduation.
Brand:  Hirschmann™ 1190133
 View more versions of this product
Product Code. 10482081
  • 24.55 € / Each
Estimated Shipment: 22-10-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Graduated to tip
  • Available in a range of capacities
Specifications
Specifications
Yes
Amber graduations permanently with silver diffusion color
1 mL
Graduated Pipet
Amber
1 Each
335 mm