Learn More
Thermo Scientific™ Graduated Safelock Microcentrifuge Tubes
GREENER_CHOICE
Easily confirm your microcentrifuge tube is locked for superior security during centrifugation.
Brand:  Thermo Scientific™ 3455IW
44.99 EUR valid until 2025-12-31
Use promo code "24328" to get your promotional price.
 View more versions of this product
Product Code. 11838332
  • 52.45 € / Pack of 500
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Find the security of a screw-cap tube with the ease-of-use of a snap-cap tube with Thermo Scientific™ Graduated Safelock Microcentrifuge Tubes. These tubes provide superior security during centrifugation. Simply position the hinged locking ball into the groove to prevent accidental cap opening, avoiding evaporation and ensuring sample integrity.
Safelock Lid Design
  • Simple one-hand operation snaps the hinged cap closed and slides the locking ball into place safeguarding valuable samples
  • Hinged locking ball reduces stress on the thumb
  • Smooth locking surface reduces the possibility of ripped or torn gloves
  • Graduation marks on the microcentrifuge tube allow for effective sample reference
Highest Quality
  • Manufactured from virgin polypropylene these microcentrifuge tubes ensure sample integrity and security during boiling, freezing, incubation and centrifugation
  • Exceptionally stable performance under the stress of centrifugation preventing tube breakage
  • Sterile product is certified free of RNase, DNase, DNA, ATP, bioburden and endotoxins
  • Nonsterile product is certified free of RNase, DNase, DNA and pyrogens
Packaging  Options
  • A variety of packaging options are available to help with contamination control in the lab
  • For stringent sterility requirements individually wrapped sterile tubes are available
Recommended For:
Daily and rigorous laboratory usage including centrifugation, boiling, incubation, freezing or sample storage.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Certified free of RNase, DNase, Pyrogen, Bioburden, ATP, PCR Inhibitors, Endotoxin and DNA.
Natural
500 Tubes/Bag, 5 Bags/Cs.
Yes
0.6 mL
Polypropylene
Sterile
30,000 x g
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Graduated Safelock Microcentrifuge Tubes >