Learn More
Deutsch Neumann™ Grey Rubber Spacers for Filtering (GUKO)
Excellent for general laboratory use. Deutsch Neumann™ Grey Rubber Spacers for Filtering (GUKO) are manufactured from natural rubber.
Brand:  Deutsch Neumann™ 2010171
 View more versions of this product
Product Code. 10420621
  • 10.15 € / Each
Estimated Shipment: 07-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Durometer 40 Shore A
  • Excellent cold flexibility
  • High tensile strength and resilience
Specifications
Specifications
Gray
1 Set
35 mm
58 mm
Natural Rubber
Spacers
78 mm