missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Deutsch Neumann™ Grey Rubber Spacers for Filtering (GUKO)
Excellent for general laboratory use. Deutsch Neumann™ Grey Rubber Spacers for Filtering (GUKO) are manufactured from natural rubber.
1.35 € - 13.30 €
Product Code | Brand | Diameter (Metric) Bottom | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Bottom | Price | Quantity & Availability | |||||
10068010
|
Deutsch Neumann™
2010051 |
53 mm |
1.35 €
Each |
Estimated Shipment: 02-01-2025
Log in to see stock. |
|||||
10420621
|
Deutsch Neumann™
2010171 |
78 mm |
10.15 €
Each |
Estimated Shipment: 06-01-2025
Log in to see stock. |
|||||
10119760
|
Deutsch Neumann™
2010181 |
89 mm |
13.30 €
Each |
Estimated Shipment: 20-12-2024
Log in to see stock. |
|||||
Description
- Durometer 40 Shore A
- Excellent cold flexibility
- High tensile strength and resilience
Specifications
Gray | |
Natural Rubber | |
Spacers |