missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark™ WypAll™ Cleaning Wiper Refill, Green

Cleans and protects with a rough side to scrub off dirt and a smooth side to clean and finish. WypAll™ Cleaning Wipers Refill is supplied pre-soaked.

Brand:  Kimberly-Clark™ 7772

127.39 EUR valid until 2024-12-31
Use promo code "24046" to get your promotional price.


 View more versions of this product

Product Code. 10760593

  • 341.00 € / Pack of 6
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Green, pre-soaked wipers
  • Rough side scrubs while smooth side polishes
  • Ideal for cleaning hands, tools, machinery without soap or water
  • Protects hands and reduces skin dryness
  • Refills for use with WypAll™ Cleaning Wiper Dispenser
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Heavy-duty Wiper
270 mm
270 mm
3.589 kg (7.91 lb.)
270 x 270 mm
For cleaning hands, tools, machinery without soap or water, protecting hands, reducing skin dryness, improving hygiene
Non-sterile
10.6 in.
10.6 in.
Paper
Green
Sheet
6 Tubs x 50 Sheets
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.