Learn More
Gilac™ Grid
Grid is highly durable and shock resistant; it avoids any contact between food and exudates.
Brand:  Gilac™ G100321
 View more versions of this product
Product Code. 17948561
  • 6.40 € / Each
Explore more special offers
Description
Description
  • Grid for flat trays.
  • Highly durable and shock resistant.
  • Enables the use of flat dishes as defrosting trays by separating the ice water from the product being defrosted.
Specifications
Specifications
Container Grid
Polyethylene
10 L Flat Tray