missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPEX™ SamplePrep Grinding Vial Accessory Packages
Contains everything necessary to run Small/Mid-size/Large grinding vials in the LargeFreezer/Mill™
1050.00 € - 1155.00 €
Specifications
Product Type | Grinding Set |
---|---|
For Use With (Equipment) | SamplePrep™ Grinding Vial, Vial Set |
Product Code | Brand | For Use With (Equipment) | Includes | Size | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Includes | Size | Price | Quantity & Availability | |||
13476148
|
SPEX™ SamplePrep
6870L |
SamplePrep™ Grinding Vial, Vial Set | 1 Vial Extractor/Opener (04576-38) and Vial Rack (04576-39) | Large |
1050.00 €
Each |
In Stock
Log in to see stock. |
|||
15330502
|
SPEX™ SamplePrep
6870S |
SamplePrep™ Grinding Vial, Vial Set | 1 Multivial Holder (04500-51), 1 Vial Extractor/Opener (04576-31), and 1 Vial Rack (04576-32) | Small |
1155.00 €
1 set |
In Stock
Log in to see stock. |
|||
Description
Small Accessory Package
- Recommended when using small grinding vials in the Large Freezer/Mill
- Compatible with: 6751 Small grinding vial set, 6761 small poly-vial set, 6771 Small low-chrome grinding vial set
- Recommended when using mid-size grinding vials in the Large Freezer/Mill
- Compatible with: 6881 mid-size grinding vial set, 6883 mid-size low-chrome grinding vial set, 6885 mid-size poly-vial set
- Recommended when using large grinding vials in the Large Freezer/Mill
- Compatible with: 6801 Large grinding vial set, 6871 Large low-chrome grinding vial set
Specifications
Grinding Set | |
SamplePrep™ Grinding Vial, Vial Set |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title