Learn More
Microspec™ Grip Seal Bags
Pack of 1000 Grip Seal Bags
Brand:  Microspec™ W5
 View more versions of this product
Product Code. 16655482
  • 41.55 € / Pack of 1000
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Grip Seal Bag
  • Sterile
  • Pack of 1000
Specifications
Specifications
Not Autoclavable
Single-use
Sterile
1000
EtO
Clear
Sample Transport
LDPE
100
2 x 9 in.