Learn More
Microspec™ Grip Seal Bags
Pack of 1000 Grip Seal Bags
Brand: Microspec™ W7
Description
- Grip Seal Bag
- Sterile
- Pack of 1000
Specifications
Not Autoclavable | |
Single-use | |
Sterile | |
1000 | |
EtO |
Clear | |
Sample Transport | |
LDPE | |
100 | |
6 x 9 in. |
Product Content Correction
Product Title
Microspec™ Grip Seal Bags > 6 x 9 in.
Spot an opportunity for improvement?Share a Content Correction