Learn More
Microspec™ Grip Seal Bags
Pack of 1000 Grip Seal Bags
Brand:  Microspec™ W8
 View more versions of this product
Product Code. 16685482
  • 73.60 € / Pack of 1000
Estimated Shipment: 18-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Grip Seal Bag
  • Sterile
  • Pack of 1000
Specifications
Specifications
Not Autoclavable
Sample Transport
LDPE
100
7.5 x 7.5 in.
Single-use
Sterile
1000
EtO