missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Notrax Gripper Sof-Tred™ Dyna-Shield™ Layer with Lengthwise Corrugated Rib Pattern
Notrax Gripper Sof-Tred™ - Exceptional worker comfort due to 12.7 mm thick specially formulated compound. Protective layer of exclusive Dyna-Shield™ extends service life by 50% compared to regular microcellular vinyl.
52.90 € - 143.00 €
Specifications
Color | Black/Yellow |
---|---|
Material | Microcellular Vinyl |
Type | Industrial and Commercial Matting |
Product Code | Brand | Length (Metric) | Width (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Width (Metric) | Price | Quantity & Availability | ||||
17742098
|
Notrax
413S0023BY |
91 cm | 60 cm |
52.90 €
Each |
Estimated Shipment: 01-10-2024
Log in to see stock. |
||||
17762098
|
Notrax
413S0035BY |
150 cm | 91 cm |
142.00 €
Each |
Estimated Shipment: 01-10-2024
Log in to see stock. |
||||
17782088
|
Notrax
413C0024BY |
1830 cm | 60 cm | ||||||
17702098
|
Notrax
413C0036BY |
1830 cm | 91 cm | ||||||
17722098
|
Notrax
413C0048BY |
1830 cm | 122 cm | ||||||
Description
- Lengthwise corrugated rib pattern for sure footing
- Exceptional worker comfort due to 127 mm thick specially formulated compound
- Protective layer of exclusive Dyna-Shield™ extends service life by 50% compared to regular microcellular vinyl
- Available with yellow borders along sides comply with OSHA code 1910-144
- All 4 sides are bevelled and sealed to minimize tripping hazards
Specifications
Black/Yellow | |
Microcellular Vinyl | |
Industrial and Commercial Matting |