missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Microscope Slides with Ground Edges

Wrapped in cellophane for improved cleanliness. These Microscope Slides with Ground Edges are washed and polished glass slides with 90° ground edge.

Brand:  Fisherbrand™ MBG01012

 View more versions of this product

Product Code. 11572203

  • 8.30 € / Pack of 50
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Glass 76 mm x 26 mm
  • Available in fully clear glass or frosted at one end
  • Ground on all four edges
Specifications

Specifications

76 mm
90°, Ground
Glass
1 to 1.2 mm
Clear
26 mm
Twin Frosted
Yes
Microscope Slide
50 Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.