missing translation for 'onlineSavingsMsg'
Learn More

Brand™ Ground Joint Sleeves

Manufactured from PTFE. BRAND™ Ground Joint Sleeves fit perfectly without a protruding rim to simplify use of joint clips. Greaseless seal will not contaminate sample. Outstanding chemical resistance at operating temperatures from -200 to 260°C.

Brand:  Brand™ 51422

28.44 EUR valid until 2025-06-13
Use promo code "24123" to get your promotional price.


Product Code. 12055332

  • 32.30 € / Pack of 10
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Manufactured from PTFE.
  • Greaseless seal will not contaminate sample.
  • Outstanding chemical resistance at operating temperatures from -200 to 260°C.
  • Thin walled sleeves (0.05mm thick) fit snugly without adding bulk.
  • Joints fit without a protruding rim to simplify use of joint clips.
  • Wide range of applications, including vacuum to 0.1mbar for rotary evaporators.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Joint clips
Ground Joint Sleeves
–200 to 260°C
NS 29/32
PTFE
10Pack
0.05mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Brand™ Ground Joint Sleeves > Size: NS 29/32

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.