missing translation for 'onlineSavingsMsg'
Learn More

Cytiva GSTrap™ 4B Prepacked Columns: 16 x 25mm

For fast and simple purification of GST-tagged proteins

Brand:  Cytiva 28-4017-48

1223.60 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.



 View more versions of this product

Product Code. 11575125

  • 1288.00 € / Pack of 5
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Prepacked with Glutathione Sepharose™ 4B media, featuring a glutathione ligand coupled to 4% agarose
  • May be used with syringe, peristaltic pump or ¨A¨KTA design chromatography system
  • Purifies GST-tagged proteins from bacterial lysates and other glutathione S-transferases or glutathione-dependent proteins
  • Bed dimensions: 16 x 25mm
  • Flow velocity: <20mL/min.
  • Average particle size: 90μm
Media
  • Matrix: Highly cross-linked 4% agarose
  • Binding capacity: >25mg horse liver GST/mL
  • pH stability working range: 4 to 13

TRUSTED_SUSTAINABILITY
Specifications

Specifications

5 mL
16 mm
<20 mL/min.
4 to 13
4°C to 30°C, 20% Ethanol
45 to 165 μm
GSTrap 4B columns are used for the purification of GST-tagged proteins from bacterial lysates and other glutathione S-tranferases or glutathione-dependent proteins. Mild elution conditions preserve protein antigenicity and function
Glutathione
0.62 in.
GSTrap 4B columns are prepacked Glutathione Sepharose 4B columns for convenient, high capacity one-step purification of glutathione S-transferase (GST) tagged proteins.
Polypropylene
4% Highly Cross-linked Agarose
5 bar (0.5 MPa, 70 psi)
5 x 5 mL
Pre-packed Column
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • GSTrap 4B 5 ml, 5 x 5 ml

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.