missing translation for 'onlineSavingsMsg'
Learn More

Cytiva GSTrap™ 4B Prepacked Columns: 16 x 25mm

For fast and simple purification of GST-tagged proteins

324.00 € - 1288.00 €

Specifications

Bed Volume 5 mL
Diameter (English) Inner 0.62 in.
Diameter (Metric) Inner 16 mm
Description GSTrap 4B columns are prepacked Glutathione Sepharose 4B columns for convenient, high capacity one-step purification of glutathione S-transferase (GST) tagged proteins.
Flow Rate <20 mL/min.
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11575125
View Documents
Cytiva
28-4017-48
5 x 5 mL
1288.00 €
Pack of 5
In Stock
to see stock.
Add to basket
 
11743319
View Documents
Cytiva
28-4017-47
1 x 5 mL
324.00 €
Each
In Stock
to see stock.
Add to basket
 
Description

Description

  • Prepacked with Glutathione Sepharose™ 4B media, featuring a glutathione ligand coupled to 4% agarose
  • May be used with syringe, peristaltic pump or ¨A¨KTA design chromatography system
  • Purifies GST-tagged proteins from bacterial lysates and other glutathione S-transferases or glutathione-dependent proteins
  • Bed dimensions: 16 x 25mm
  • Flow velocity: <20mL/min.
  • Average particle size: 90μm
Media
  • Matrix: Highly cross-linked 4% agarose
  • Binding capacity: >25mg horse liver GST/mL
  • pH stability working range: 4 to 13

Specifications

Specifications

5 mL
16 mm
<20 mL/min.
4 to 13
4°C to 30°C, 20% Ethanol
45 to 165 μm
Pre-packed Column
0.62 in.
GSTrap 4B columns are prepacked Glutathione Sepharose 4B columns for convenient, high capacity one-step purification of glutathione S-transferase (GST) tagged proteins.
Polypropylene
4% Highly Cross-linked Agarose
5 bar (0.5 MPa, 70 psi)
GSTrap 4B columns are used for the purification of GST-tagged proteins from bacterial lysates and other glutathione S-tranferases or glutathione-dependent proteins. Mild elution conditions preserve protein antigenicity and function
Glutathione
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • GSTrap 4B 5 ml, 5 x 5 ml

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.