missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva GSTrap™ 4B Prepacked Columns: 16 x 25mm
For fast and simple purification of GST-tagged proteins
Brand: Cytiva 28-4017-47
307.80 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.
This item is not returnable.
View return policy
Description
- Prepacked with Glutathione Sepharose™ 4B media, featuring a glutathione ligand coupled to 4% agarose
- May be used with syringe, peristaltic pump or ¨A¨KTA design chromatography system
- Purifies GST-tagged proteins from bacterial lysates and other glutathione S-transferases or glutathione-dependent proteins
- Bed dimensions: 16 x 25mm
- Flow velocity: <20mL/min.
- Average particle size: 90μm
- Matrix: Highly cross-linked 4% agarose
- Binding capacity: >25mg horse liver GST/mL
- pH stability working range: 4 to 13
Specifications
5 mL | |
16 mm | |
<20 mL/min. | |
4 to 13 | |
4°C to 30°C, 20% Ethanol | |
45 to 165 μm | |
GSTrap 4B columns are used for the purification of GST-tagged proteins from bacterial lysates and other glutathione S-tranferases or glutathione-dependent proteins. Mild elution conditions preserve protein antigenicity and function | |
Glutathione |
0.62 in. | |
GSTrap 4B columns are prepacked Glutathione Sepharose 4B columns for convenient, high capacity one-step purification of glutathione S-transferase (GST) tagged proteins. | |
Polypropylene | |
4% Highly Cross-linked Agarose | |
5 bar (0.5 MPa, 70 psi) | |
1 x 5 mL | |
Pre-packed Column |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- GSTrap 4B 5 ml, 1 x 5 ml
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction