missing translation for 'onlineSavingsMsg'
Learn More
Guanethidine sulfate, Thermo Scientific™ Product Code.: 17312491

Guanethidine sulfate, Thermo Scientific™

Product Code. 17312491
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 17312491

Brand: Thermo Scientific Acros 468620010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 645-43-2
Molecular Formula C10H24N4O4S
Molecular Weight (g/mol) 296.39
MDL Number MFCD00035403
InChI Key YUFWAVFNITUSHI-UHFFFAOYSA-N
IUPAC Name N''-[2-(azocan-1-yl)ethyl]guanidine; sulfuric acid
SMILES OS(O)(=O)=O.NC(N)=NCCN1CCCCCCC1

Specifications

Packaging Glass Bottle
Quantity 1 g
Certificate of Analysis Available
Concentration or Composition Notes Titration with HClO4
Physical Form Crystalline Powder/Crystals
Concentration 97.5% to 102.5%
Color White to Light Beige
Formula Weight 296.39
Chemical Name or Material Guanethidine sulfate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0458379 Certificate TypeCertificate of Analysis Date11/12/2023 Catalog Number468622500,468620010
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Guanethidine sulfate, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.