missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
Packaging | Glass Bottle |
Quantity | 250 mg |
Certificate of Analysis | Available |
Concentration or Composition Notes | Titration with HClO4 |
Physical Form | Crystalline Powder/Crystals |
Concentration | 97.5% to 102.5% |
Color | White to Light Beige |
Formula Weight | 296.39 |
Chemical Name or Material | Guanethidine sulfate |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0458379 | Certificate of Analysis | 11/12/2023 | 468622500,468620010 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Guanethidine sulfate, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction