missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OHAUS™ Guardian 5000 Hotplate-Stirrer
Reliable hotplate stirrers with exceptional safety features for everyday use.
610.00 € - 979.00 €
Product Code | Brand | Model | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Price | Quantity & Availability | |||||
16640452
|
OHAUS™
30500523 |
e-G51HS07C | N/A | ||||||
16670452
|
OHAUS™
30500553 |
e-G51HS10C |
979.00 €
Each |
In Stock
Log in to see stock. |
|||||
16680452
|
OHAUS™
30500563 |
e-G51HSRDM |
610.00 €
Each |
In Stock
Log in to see stock. |
|||||
Description
- An industry-leading early detection system that uses two independent safety controls to continuously monitor the electronics and shut off heating before an overtemperature condition occurs.
- Precise stirring from 60 to 1600 rpm is made possible by the design of a powerful motor, strong magnet and software controlled ramp rate, ensuring secure magnetic coupling for viscous applications.
- The bright LCD display includes large temperature and speed settings, intuitive icons and a prominent hot top indicator - all designed to quickly and easliy monitor performance from across the lab.
Specifications
230 V | |
50/60 Hz | |
EU |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title