Learn More
Tocris Bioscience™ GYY 4137 morpholine salt
Slow-releasing H2S donor
Brand:  Tocris Bioscience™ 3658/10
101.82 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18085902
Description
Description
GYY 4137 morpholine salt is a slow-releasing H2S donor. Exhibits vasodilator and antihypertensive activity. Activity causes slow dilation of blood vessels in vitro and in vivo; does not influence vascular smooth muscle cell viability in culture. Water-soluble.
Specifications
Specifications
10 mg
GYY 4137 morpholine salt
376.47
GYY 4137 morpholine salt