Learn More
Tocris Bioscience™ GYY 4137 morpholine salt
Slow-releasing H2S donor
127.00 € - 434.00 €
Specifications
Product Type | GYY 4137 morpholine salt |
---|---|
Molecular Weight (g/mol) | 376.47 |
Inhibitors | GYY 4137 morpholine salt |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18085902
|
Tocris Bioscience™
3658/10 |
10 mg |
127.00 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18093912
|
Tocris Bioscience™
3658/50 |
50 mg |
434.00 €
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Slow-releasing H2S donor. Exhibits vasodilator and antihypertensive activity. Activity causes slow dilation of blood vessels in vitro and in vivo; does not influence vascular smooth muscle cell viability in culture. Water-soluble.Specifications
GYY 4137 morpholine salt | |
376.47 | |
GYY 4137 morpholine salt |