Learn More
SP Bel-Art™ H-B Durac Plus Oven Verification Thermometer
These H-B Durac Plus Oven Verification Thermometers are mercury free and traceable to NIST
Brand:  SP Bel-Art™ B60600-1300
 View more versions of this product
Product Code. 17561285
  • 67.25 € / Each
Explore more special offers
Description
Description
  • Verify sample temperatures in ovens
  • Accurate temperature reading without sample contamination is achieved by using the filled bottle (included), which mimics the sample
  • Insert the thermometer into the bottle and place near the samples for best results
  • Thermometers contain organic liquid, or EnviroKleen™ certified Enviro-Safe™ liquid, and feature permanently fused markings and lead free glass
  • PFA coating on select thermometers safely contain glass and liquid in case of breakage
  • Manufactured and tested by our USA quality team; individually serialized
  • Includes multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability
  • Should you require an individual Thermometer Calibration Report, factory calibrated thermometers or custom calibration services are available
Specifications
Specifications
Glass Beads
145 mm
Thermometer
50°C to 200°C
30 mL
Organic Liquid
5.7 in.