missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SP Bel-Art™ H-B Durac Plus Oven Verification Thermometer
These H-B Durac Plus Oven Verification Thermometers are mercury free and traceable to NIST
67.25 € - 68.05 €
Specifications
Bottle Fill | Glass Beads |
---|---|
Bottle Size | 30 mL |
Thermometer Fill | Organic Liquid |
Product Type | Thermometer |
Product Code | Brand | Temperature Range (Metric) | Accuracy | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Temperature Range (Metric) | Accuracy | Price | Quantity & Availability | ||||
17551285
|
SP Bel-Art™
B60600-1100 |
20°C to 130°C | ±1.5°C; ±2°C above 100°C |
68.05 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17561285
|
SP Bel-Art™
B60600-1300 |
50°C to 200°C |
67.25 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Verify sample temperatures in ovens
- Accurate temperature reading without sample contamination is achieved by using the filled bottle (included), which mimics the sample
- Insert the thermometer into the bottle and place near the samples for best results
- Thermometers contain organic liquid, or EnviroKleen™ certified Enviro-Safe™ liquid, and feature permanently fused markings and lead free glass
- PFA coating on select thermometers safely contain glass and liquid in case of breakage
- Manufactured and tested by our USA quality team; individually serialized
- Includes multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability
- Should you require an individual Thermometer Calibration Report, factory calibrated thermometers or custom calibration services are available
Specifications
Glass Beads | |
30 mL | |
Organic Liquid | |
Thermometer |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P370+P378-In case of fire: Use to extinguish.
- P403+P235-Store in a well-ventilated place. Keep cool.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title