Learn More
H-B Instrument™ Durac™ Compact Sling Psychrometer
Measures relative humidity with weather bureau-type wet and dry bulb.
Brand:  H-B Instrument™ B61503-0000
Product Code. 17590685
  • 105.28 € / Each
Explore more special offers
Description
Description
  • Organic liquid filled thermometers with white back glass are mounted in a pocket sized self contained plastic case designed with a built in slide rule/convert wet and dry bulb readings/relative humidity, thus eliminating the need for calculating tables
Specifications
Specifications
Compact Sling Psychrometer
-3.88°C to +50°C
Glass
8.3 in.
To measure relative humidity with weather bureau-type wet and dry bulb
1 ea.
1°C
23°F to 122°F
21 cm
White back glass
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
H-B Instrument™ Durac™ Compact Sling Psychrometer > Qty.: 5 pieces