missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ HAAKE™ Viscotester™ 3 Rotational Viscometer

Ingenious design makes this portable viscometer the easiest you’ll ever use for fast, reliable measurements anywhere in your process.

3175.00 €

Specifications

Rotational Speed Range 62.5 rpm
Temperature Range (Metric) Up to 150°C
View More Specs

Products 1
Product Code Brand Description Viscosity Range Price Quantity & Availability  
Product Code Brand Description Viscosity Range Price Quantity & Availability  
16687305
View Documents
Thermo Scientific™
399-0112
HAAKE Viscotester 3 Rotational Viscometer for higher viscosity measurements 30 to 400 000 mPas
3175.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Perform viscosity measurements with ease. The Thermo Scientific™ HAAKE™ Viscotester™ 3 rotational viscometer ensures quick, reliable tests for QC applications.

Use it as a handheld viscometer or on a lab stand. Color coding ensures measuring geometries match the analysis, the display is readable in any lighting, and the viscometer remembers the reference point measurement for you. All that comes in a ready-to-go carrying case.

HAAKE Viscotester 3 Rotational Viscometer is the replacement for the discontinued Thermo Scientific™ HAAKE™ Viscotester™ 1 plus and 2 plus Rotational Viscometers.

Designed for simplicity – The HAAKE Viscotester 3 Rotational Viscometer is built for ease of use to get you quickly to the right viscosity measurements:
  • Smart alignment given by design and visualized by an electronic level
  • Quick coupling of measuring geometries for fast and easy adaptation
  • Color coded geometries and a geometry range indicator ensure correct rotor selection every time
  • Adaptable display brightness ensures ideal readability in any environment
  • Battery and power supply options allow the viscometer to be run anywhere
  • Memory assist functionality allows you to quickly and easily compare measured viscosities with the reference point
  • Selectable viscosity units (mPa s or dPa s)
  • Compatibility with previous viscometer models

Recommended for: your entire process, from accepting the delivery of goods, to sampling during production and quality control (QC).
  • Printing inks, paints, inks and other coatings
  • Shampoos, creams, lotions, and similar personal care products
  • Oil, greases, pastes and similar substances
  • Sauces, thickeners, and similar food products
Specifications

Specifications

62.5 rpm
Up to 150°C
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.