Learn More
Gilac™ HACCP Flat Tray
HACCP Flat Tray can be used in a wide range of temperatures and its dimensions are suitable for refrigerators.
10.05 € - 18.40 €
Product Code | Brand | Capacity (Metric) | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Color | Price | Quantity & Availability | |||||
17928961
|
Gilac™
G155221 |
3 L | White |
10.05 €
Each |
Estimated Shipment: 09-05-2025
Log in to see stock. |
|||||
17928821
|
Gilac™
G155210 |
3 L | Blue |
10.05 €
Each |
Estimated Shipment: 09-05-2025
Log in to see stock. |
|||||
17988401
|
Gilac™
G155211 |
3 L | Green |
10.05 €
Each |
Estimated Shipment: 09-05-2025
Log in to see stock. |
|||||
17908601
|
Gilac™
G155209 |
3 L | Red |
10.05 €
Each |
Estimated Shipment: 09-05-2025
Log in to see stock. |
|||||
17948711
|
Gilac™
G155218 |
3 L | Yellow |
10.05 €
Each |
Estimated Shipment: 09-05-2025
Log in to see stock. |
|||||
17908641
|
Gilac™
G155621 |
5 L | White |
14.15 €
Each |
Estimated Shipment: 09-05-2025
Log in to see stock. |
|||||
17968541
|
Gilac™
G155710 |
8 L | Blue |
18.40 €
Each |
Estimated Shipment: 09-05-2025
Log in to see stock. |
|||||
17908971
|
Gilac™
G155711 |
8 L | Green |
18.40 €
Each |
Estimated Shipment: 09-05-2025
Log in to see stock. |
|||||
17908481
|
Gilac™
G155718 |
8 L | Yellow |
18.40 €
Each |
Estimated Shipment: 09-05-2025
Log in to see stock. |
|||||
17918421
|
Gilac™
G155721 |
8 L | White |
18.40 €
Each |
Estimated Shipment: 09-05-2025
Log in to see stock. |
|||||
Description
- Flat trays.
- Large and customisable range.
- 5 colors.
- 5 volumes.
- Can be used in a wide range of temperatures.
- Dimensions suitable for refrigerators.
- Can be used as a:
- defrosting tray thanks to its wide dividing grid that separates the ice water from the product being defrosted.
- preparation container for products that drip.
- ingredients tray.
- container for preserving comparison samples.
- Fermenting and decanting containers.
- Nesting and stacking with lid.
- Traceability management thanks (IML) injection molding.
Specifications
HDPE | |
Dispensing Tray | |
Not Autoclavable |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Gilac™ HACCP Flat Tray