Learn More
Gilac™ HACCP Food Contact Container
HACCP Food Contact Container is ideal for storage, food preparation, and transporting foodstuffs.
Brand:  Gilac™ G613233
 View more versions of this product
Product Code. 17968911
  • 47.75 € / Each
Explore more special offers
Description
Description
  • HACCP round container.
  • Food contact certified.
  • For storage, food preparation and transporting foodstuffs.
  • Highly durable.
  • Reinforced bottom with anti-wear skirt.
  • Reinforced handles to prevent distortion during carrying.
  • Optional, choice of two lids.
  • External Diameter includes handles.
  • Width corresponds to diameter without handles.
Specifications
Specifications
HACCP Food Container
Red
50 L
Polyethylene
13.2 gal.