missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HAL Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA529123
This item is not returnable.
View return policy
Description
Recommended positive controls: mouse liver. Predicted reactivity: Mouse (96%), Rat (96%), Zebrafish (90%), Chicken (94%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
HAL is a cytosolic enzyme catalyzing the first reaction in histidine catabolism, the nonoxidative deamination of L-histidine to trans-urocanic acid. HAL defects cause histidinemia which is characterized by increased histidine and histamine and decreased urocanic acid in body fluids Histidine ammonia-lyase is a cytosolic enzyme catalyzing the first reaction in histidine catabolism, the nonoxidative deamination of L-histidine to trans-urocanic acid. Histidine ammonia-lyase defects cause histidinemia which is characterized by increased histidine and histamine and decreased urocanic acid in body fluids.
Specifications
HAL | |
Polyclonal | |
Unconjugated | |
HAL | |
hal; HIS; histidase; histidine ammonia lyase; histidine ammonia-lyase; histidinemia; Hsd; HSTD; Huth; zgc:136980 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
15109, 3034 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
P35492, P42357 | |
HAL | |
Recombinant fragment corresponding to a region within amino acids 209 and 489 of Human HAL | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
HAL Polyclonal Antibody, Invitrogen™ > 100 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction