Learn More
CleanSpace™ Half Mask Adaptor
Half mask adaptor is equipped for the front of the mask to enable quantitative fit testing.
Brand:  CleanSpace™ PAF-0025
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15950052
  • 251.00 € / Each
Estimated Shipment: 19-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Used with the revolutionary CleanSpace™ Respirators: light weight, no hoses or belts
  • Used with the CleanSpace™ Half Masks only
  • Quick and easy to install
  • Fits with the TSI PortaCount equipment
  • Easy to wash and quick drying
Specifications
Specifications
Half Mask Adapter
Quantitative Fit Testing
PAF-1010, PAF-0027, PAF-0033
PAF-0025
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
CleanSpace™ Half Mask Adaptor > Product Type: Half Mask Adaptor